Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn7l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240009.2 | XP_006240071.1 | 2 | 272 | ||
XM_006240015.2 | XP_006240077.1 | 2 | 272 | |||
XM_006240016.2 | XP_006240078.1 | 2 | 272 | |||
XM_006240019.2 | XP_006240081.1 | 2 | 272 | |||
XM_008764632.1 | XP_008762854.1 | 2 | 272 | |||
XM_008764634.1 | XP_008762856.1 | 2 | 272 | |||
GenBank mRNA | BC166538.1 | - | 2 | 154 |