Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100903.1 | NP_001094373.1 | 2 | 131 | ||
XM_006241072.2 | XP_006241134.1 | 2 | 135 | |||
XM_006241073.2 | XP_006241135.1 | 2 | 339 | |||
XM_006241074.2 | XP_006241136.1 | 2 | 1063 | |||
XM_008765170.1 | XP_008763392.1 | 1 | 1854 | |||
XM_008765171.1 | XP_008763393.1 | 1 | 1854 | |||
XM_008765172.1 | XP_008763394.1 | 1 | 1854 | |||
GenBank mRNA | BC100641.1 | - | 2 | 78 |