Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108783.1 | NP_001102253.1 | 6 | 428 | ||
XM_006243970.2 | XP_006244032.1 | 7 | 1052 |