Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108788.1 | NP_001102258.1 | 12 | 1633 | ||
XM_006244461.2 | XP_006244523.1 | 13 | 1704 | |||
XM_006244462.2 | XP_006244524.1 | 12 | 1660 | |||
XM_008766866.1 | XP_008765088.1 | 13 | 1664 | |||
XM_008766867.1 | XP_008765089.1 | 13 | 1661 | |||
XM_008766868.1 | XP_008765090.1 | 13 | 1643 | |||
XM_008766869.1 | XP_008765091.1 | 13 | 1607 | |||
XM_008766870.1 | XP_008765092.1 | 13 | 1604 | |||
XM_008766871.1 | XP_008765093.1 | 13 | 1640 |