Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other POU4F3 SIRNA Assays ›
Gene Symbol:
Pou4f3
Gene Name:
POU class 4 homeobox 3
Gene Aliases:
-
Chromosome Location:
Chr.18: 36713869 - 36716461 on Build Rnor_6.0
UniGene ID:
Rn.218181
Species:
Rat
siRNA ID
RSS338710
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Pou4f3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001108889.1
NP_001102359.1
2
863
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
sensory perception of sound
biological_process
vestibulocochlear nerve development
retinal ganglion cell axon guidance
inner ear morphogenesis
auditory receptor cell differentiation
positive regulation of transcription from RNA polymerase II promoter
axon extension
inner ear development
neuromuscular process controlling balance
neuron apoptotic process
inner ear receptor cell differentiation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
molecular_function
DNA binding
transcription factor activity, sequence-specific DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-69b65fdbbd-2sdz6:80/100.66.76.100:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline