Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000694.1 | NP_001000694.1 | 70 |
Gene Symbol: LOC100910009 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748917.2 | XP_003748965.1 | 1 | 146 |
Gene Symbol: Olr294 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748914.2 | XP_003748962.1 | 146 |