Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sftpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017342.2 | NP_059038.1 | 2, 3 | 219 | ||
GenBank mRNA | BC072693.1 | - | 2, 3 | 212 | 7099876 | |
X14221.1 | - | 2, 3 | 220 |