Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019292.4 | NP_062165.2 | 6, 7 | 661 | ||
XM_006232116.1 | XP_006232178.1 | 7, 8 | 986 | |||
GenBank mRNA | AB030829.1 | - | 6, 7 | 661 | ||
AB030830.1 | - | 6, 7 | 661 | |||
AF037072.1 | - | 6, 7 | 684 | |||
BC061980.1 | - | 6, 7 | 696 | 5597975 | ||
FQ210528.1 | - | 6, 7 | 709 | |||
FQ211459.1 | - | 6, 7 | 708 | |||
FQ214846.1 | - | 6, 7 | 706 | |||
FQ214929.1 | - | 6, 7 | 711 | |||
FQ215392.1 | - | 5, 6 | 523 | |||
FQ216182.1 | - | 6, 7 | 707 | |||
FQ223846.1 | - | 6, 7 | 707 | |||
M22413.1 | - | 6, 7 | 816 |