Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019328.3 | NP_062201.2 | 2 | 203 | ||
XM_006234201.2 | XP_006234263.1 | 3 | 421 | |||
GenBank mRNA | DQ092629.1 | - | 2 | 134 | ||
DQ092631.1 | - | 2 | 148 | |||
DQ092632.1 | - | 2 | 138 | |||
FQ222109.1 | - | 3 | 442 | |||
L08595.1 | - | 2 | 203 | |||
U01146.1 | - | 2 | 134 | |||
U72345.1 | - | 3 | 427 |