Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020092.1 | NP_064477.1 | 4, 5 | 554 | ||
XM_006231892.2 | XP_006231954.1 | 6, 7 | 1073 | |||
XM_006231894.2 | XP_006231956.1 | 5, 6 | 5482 | |||
GenBank mRNA | AF234765.1 | - | 4, 5 | 554 |