Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crhr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247542.2 | XP_006247604.1 | 6 | 1883 | ||
GenBank mRNA | EU012436.1 | - | 7 | 609 | ||
EU012438.1 | - | 7 | 677 | |||
L24096.1 | - | 7 | 690 | |||
L25438.1 | - | 7 | 612 |
Set Membership: |
G-protein coupled receptor |