Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2k2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133283.1 | NP_579817.1 | 4 | 540 | ||
XM_006240987.2 | XP_006241049.1 | 4 | 727 | |||
GenBank mRNA | BC126084.1 | - | 4 | 638 | 8376547 | |
D14592.1 | - | 4 | 511 | |||
L14936.1 | - | 4 | 540 |