Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021659.1 | NP_067691.1 | 7 | 1008 | ||
XM_006231065.2 | XP_006231127.1 | 11 | 2003 | |||
XM_006231066.2 | XP_006231128.1 | 10 | 2399 | |||
XM_006231067.2 | XP_006231129.1 | 10 | 1871 | |||
XM_006231068.2 | XP_006231130.1 | 10 | 1775 | |||
XM_006231069.2 | XP_006231131.1 | 8 | 1283 | |||
GenBank mRNA | AF336852.1 | - | 12 | 1823 | ||
AF336853.1 | - | 11 | 1595 | |||
AF336854.1 | - | 7 | 848 | |||
AF336855.1 | - | 11 | 1700 | |||
AF336856.1 | - | 9 | 1199 | |||
AF336857.1 | - | 9 | 1208 | |||
AF336858.1 | - | 9 | 1340 | |||
AF336859.1 | - | 10 | 1472 | |||
AF336860.1 | - | 10 | 1568 | |||
U20106.1 | - | 7 | 1008 |