Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021666.2 | NP_067698.1 | 1971 | |||
GenBank mRNA | AJ243304.2 | - | 1971 |