Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021858.3 | NP_068630.1 | 5, 6 | 375 | ||
XM_006237383.1 | XP_006237445.1 | 5, 6 | 1357 | |||
GenBank mRNA | BC086422.1 | - | 5, 6 | 378 | 7193131 | |
DQ120487.1 | - | 3, 4 | 185 | |||
DQ120488.1 | - | 3, 4 | 185 |