Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctla4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031674.1 | NP_113862.1 | 1 | 12 | ||
GenBank mRNA | U37121.1 | - | 1 | 12 | ||
U90271.1 | - | 1 | 12 |