Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270701.1 | NP_001257630.1 | 4 | 881 | ||
NM_001270702.1 | NP_001257631.1 | 4 | 794 | |||
NM_001270703.1 | NP_001257632.1 | 4 | 881 | |||
NM_023021.2 | NP_075410.2 | 4 | 881 | |||
XM_006228466.2 | XP_006228528.1 | 4 | 2641 | |||
XM_006228467.2 | XP_006228529.1 | 4 | 2642 | |||
XM_006228469.1 | XP_006228531.1 | 5 | 863 | |||
XM_006228470.2 | XP_006228532.1 | 4 | 711 | |||
XM_006228471.1 | XP_006228533.1 | 4 | 3138 | |||
XM_008758975.1 | XP_008757197.1 | 5 | 2536 | |||
XM_008758976.1 | XP_008757198.1 | 4 | 1060 | |||
XM_008758977.1 | XP_008757199.1 | 3 | 3042 | |||
XM_008758978.1 | XP_008757200.1 | 4 | 700 | |||
GenBank mRNA | AB292802.1 | - | 2 | 356 | ||
AB292803.1 | - | 4 | 739 | |||
AF149250.1 | - | 4 | 739 | |||
AF156554.1 | - | 4 | 881 | |||
AF190458.1 | - | 4 | 851 | |||
AJ133438.2 | - | 4 | 739 | |||
BC128736.1 | - | 4 | 832 | 8372655 | ||
DQ137429.1 | - | 1 | 23 | |||
EU872449.1 | - | 4 | 854 | |||
EU872450.1 | - | 4 | 854 | |||
EU872451.1 | - | 4 | 767 |