Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kiss1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301151.1 | NP_001288080.1 | 1 | 359 | ||
NM_023992.2 | NP_076482.2 | 1 | 359 | |||
GenBank mRNA | AB051066.1 | - | 1 | 185 | ||
AF115516.1 | - | 1 | 185 |
Set Membership: |
G-protein coupled receptor |