Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jup | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031047.2 | NP_112309.2 | 5, 6 | 1297 | ||
XM_006247429.1 | XP_006247491.1 | 5, 6 | 1068 | |||
XM_006247430.2 | XP_006247492.1 | 5, 6 | 1034 | |||
GenBank mRNA | BC065580.1 | - | 5, 6 | 1297 | 5599724 | |
FQ223166.1 | - | 5, 6 | 1057 | |||
U58858.1 | - | 5, 6 | 1049 |