Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031144.3 | NP_112406.1 | 4, 5 | 870 | ||
GenBank mRNA | AB028846.1 | - | 1, 2 | 79 | ||
BC063166.1 | - | 4, 5 | 873 | 6920838 | ||
EF156276.1 | - | 2, 3 | 592 | |||
FQ226945.1 | - | 144 | ||||
FQ227027.1 | - | 432 | ||||
FQ227201.1 | - | 229 | ||||
FQ229293.1 | - | 231 | ||||
FQ231277.1 | - | 226 | ||||
FQ231503.1 | - | 4, 5 | 872 | |||
FQ231691.1 | - | 220 | ||||
FQ232063.1 | - | 4, 5 | 872 | |||
FQ232650.1 | - | 4, 5 | 872 | |||
FQ232682.1 | - | 412 |