Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031777.2 | NP_113965.1 | 4 | 274 | ||
XM_006250266.2 | XP_006250328.1 | 4 | 459 | |||
GenBank mRNA | AF026476.1 | - | 2 | 143 | ||
BC088849.1 | - | 4 | 274 | 7313699 |