Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stmn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053440.2 | NP_445892.1 | 2 | 116 | ||
GenBank mRNA | AF306458.1 | - | 2 | 214 | ||
BC087660.1 | - | 2 | 116 | 7313867 |