Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco1a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130736.1 | NP_570092.1 | 11 | 1620 | ||
XM_006237650.2 | XP_006237712.1 | 12 | 1674 | |||
GenBank mRNA | AF053317.2 | - | 11 | 1620 |