Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sos2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135561.1 | NP_001129033.1 | 14 | 2279 | ||
XM_006240182.2 | XP_006240244.1 | 14 | 2418 | |||
XM_006240184.2 | XP_006240246.1 | 11 | 2824 | |||
XM_008764721.1 | XP_008762943.1 | 13 | 2322 |