Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138831.1 | NP_620186.1 | 3 | 1093 | ||
XM_008772974.1 | XP_008771196.1 | 3 | 1132 | |||
XM_008772975.1 | XP_008771197.1 | 3 | 1131 | |||
GenBank mRNA | AB047324.1 | - | 3 | 1093 |