Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133398.1 | NP_596889.1 | 8 | 1560 | ||
XM_006241520.2 | XP_006241582.1 | 9 | 3069 | |||
XM_006241521.2 | XP_006241583.1 | 8 | 3004 | |||
XM_006241523.2 | XP_006241585.1 | 7 | 2911 | |||
XM_006241524.2 | XP_006241586.1 | 7 | 2878 | |||
XM_006241525.2 | XP_006241587.1 | 6 | 2813 | |||
GenBank mRNA | AF100421.2 | - | 8 | 1560 | ||
FQ225234.1 | - | 4 | 543 | |||
FQ225997.1 | - | 4 | 543 | |||
FQ226094.1 | - | 8 | 1556 | |||
FQ227915.1 | - | 6 | 708 | |||
FQ231703.1 | - | 5 | 642 | |||
FQ233989.1 | - | 3 | 406 | |||
FQ234551.1 | - | 7 | 1498 |