Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nos1ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138922.1 | NP_620277.1 | 10 | 1661 | ||
GenBank mRNA | AF037071.1 | - | 10 | 1661 |