Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adra2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138505.2 | NP_612514.2 | 1 | 1416 | ||
GenBank mRNA | M32061.1 | - | 1 | 1413 |
Set Membership: |
G-protein coupled receptor |