Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nradd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139259.2 | NP_640352.1 | 3, 4 | 412 | ||
XM_006243921.2 | XP_006243983.1 | 3, 4 | 416 | |||
GenBank mRNA | AF497263.1 | - | 3, 4 | 416 | ||
AF497264.1 | - | 3, 4 | 264 | |||
AF534395.1 | - | 2, 3 | 228 |