Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017060.2 | NP_058756.2 | 5 | 527 | ||
XM_006230687.2 | XP_006230749.1 | 4 | 1396 | |||
GenBank mRNA | AB298805.1 | - | 3 | 446 | ||
CB794299.1 | - | 2 | 233 | |||
FQ224126.1 | - | 5 | 587 |