Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aanat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012818.2 | NP_036950.1 | 4 | 658 | ||
XM_006247792.1 | XP_006247854.1 | 4 | 1654 | |||
XM_006247793.1 | XP_006247855.1 | 4 | 1649 | |||
GenBank mRNA | DQ075321.1 | - | 4 | 846 | ||
U38306.1 | - | 4 | 658 | |||
U40803.1 | - | 4 | 745 |