Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012771.3 | NP_036903.2 | 1 | 108 | ||
GenBank mRNA | BC058490.1 | - | 1 | 112 | 6919643 | |
FQ221278.1 | - | 1 | 108 | |||
FQ223335.1 | - | 1 | 108 | |||
FQ226475.1 | - | 1 | 108 | |||
FQ226588.1 | - | 1 | 108 | |||
FQ227076.1 | - | 1 | 108 | |||
FQ227452.1 | - | 1 | 108 | |||
FQ227932.1 | - | 1 | 108 | |||
FQ228036.1 | - | 1 | 108 | |||
FQ229300.1 | - | 1 | 108 | |||
FQ230656.1 | - | 1 | 108 | |||
FQ231085.1 | - | 1 | 108 | |||
FQ231872.1 | - | 1 | 108 | |||
FQ231961.1 | - | 1 | 108 | |||
FQ233423.1 | - | 1 | 108 | |||
FQ233793.1 | - | 1 | 108 |