Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bckdha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012782.1 | NP_036914.1 | 4 | 394 | ||
GenBank mRNA | BC089915.1 | - | 2 | 41 | ||
BC161819.1 | - | 4 | 399 | 9099948 | ||
CB579195.1 | - | 4 | 400 | |||
J02827.1 | - | 4 | 378 |