Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gamt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012793.2 | NP_036925.2 | 4 | 494 | ||
GenBank mRNA | FM043834.1 | - | 4 | 467 | ||
FQ210612.1 | - | 4 | 417 | |||
FQ225896.1 | - | 4 | 494 | |||
FQ227114.1 | - | 4 | 494 | |||
J03588.1 | - | 4 | 492 |