Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkbia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105720.2 | NP_001099190.2 | 3 | 601 | ||
GenBank mRNA | BC166886.1 | - | 3 | 553 | 6890357 | |
CB316505.1 | - | 3 | 555 | |||
FQ214065.1 | - | 1 | 49 | |||
FQ215246.1 | - | 3 | 553 | |||
FQ225350.1 | - | 3 | 548 | |||
FQ226288.1 | - | 3 | 551 | |||
FQ227283.1 | - | 3 | 551 | |||
FQ233200.1 | - | 3 | 551 | |||
FQ233557.1 | - | 3 | 553 | |||
FQ233627.1 | - | 3 | 539 | |||
FQ234262.1 | - | 3 | 553 | |||
X63594.1 | - | 3 | 531 |