Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkbia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105720.2 | NP_001099190.2 | 5, 6 | 1009 | ||
GenBank mRNA | BC166886.1 | - | 5, 6 | 961 | 6890357 | |
BF417507.1 | - | 1, 2 | 131 | |||
CA509173.1 | - | 1, 2 | 572 | |||
CB324027.1 | - | 1, 2 | 572 | |||
FQ214065.1 | - | 3, 4 | 457 | |||
FQ215246.1 | - | 5, 6 | 961 | |||
FQ225350.1 | - | 5, 6 | 956 | |||
FQ225877.1 | - | 5, 6 | 959 | |||
FQ226288.1 | - | 5, 6 | 959 | |||
FQ227283.1 | - | 5, 6 | 959 | |||
FQ233200.1 | - | 5, 6 | 959 | |||
FQ233557.1 | - | 5, 6 | 961 | |||
FQ233627.1 | - | 5, 6 | 947 | |||
FQ234262.1 | - | 5, 6 | 962 | |||
X63594.1 | - | 5, 6 | 939 |