Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013112.1 | NP_037244.1 | 2 | 100 | ||
XM_006250238.2 | XP_006250300.1 | 3 | 234 | |||
XM_006250239.2 | XP_006250301.1 | 3 | 240 | |||
GenBank mRNA | FQ209753.1 | - | 3 | 165 | ||
FQ209821.1 | - | 3 | 165 | |||
FQ210214.1 | - | 3 | 158 | |||
FQ210245.1 | - | 3 | 165 | |||
FQ210295.1 | - | 3 | 162 | |||
FQ210325.1 | - | 3 | 185 | |||
FQ210458.1 | - | 3 | 162 | |||
FQ210462.1 | - | 3 | 163 | |||
FQ210537.1 | - | 2 | 159 | |||
FQ210542.1 | - | 2 | 288 | |||
FQ210648.1 | - | 3 | 162 | |||
FQ210885.1 | - | 3 | 192 | |||
FQ210979.1 | - | 3 | 165 | |||
FQ211006.1 | - | 3 | 159 | |||
FQ211265.1 | - | 3 | 162 | |||
FQ211383.1 | - | 3 | 164 | |||
FQ218273.1 | - | 3 | 165 | |||
FQ218326.1 | - | 3 | 159 | |||
FQ218756.1 | - | 3 | 166 | |||
FQ221741.1 | - | 3 | 165 | |||
FQ223965.1 | - | 3 | 159 | |||
M28615.1 | - | 2 | 69 | |||
X03468.1 | - | 2 | 100 |