Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172243.1 | NP_758443.1 | 6 | 635 | ||
GenBank mRNA | BC086977.1 | - | 6 | 668 | 7193156 | |
U68544.1 | - | 6 | 635 |