Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004202.2 | NP_001004202.1 | 1, 2 | 70 | ||
GenBank mRNA | AY620903.1 | - | 1, 2 | 80 | ||
BC079460.1 | - | 102 | 7121977 | |||
EF121992.1 | - | 1, 2 | 70 | |||
EF121993.1 | - | 1, 2 | 70 |