Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100912275 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751285.3 | XP_003751333.1 | 2 | 632 | ||
XM_008769810.1 | XP_008768032.1 | 2 | 674 |
Gene Symbol: Mixl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105979.1 | NP_001099449.1 | 2 | 654 | ||
XM_008769818.1 | XP_008768040.1 | 2 | 674 |