Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcg3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004076.2 | NP_001004076.2 | 9 | 1020 | ||
XM_006250626.2 | XP_006250688.1 | 9 | 1069 | |||
XM_008770002.1 | XP_008768224.1 | 9 | 3410 | |||
XM_008770003.1 | XP_008768225.1 | 10 | 3290 | |||
GenBank mRNA | AY688113.1 | - | 8 | 947 | ||
BC098896.1 | - | 9 | 1020 | 7100556 |