Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728408.4 | XP_002728454.2 | 3 | 1141 | ||
XM_006253172.2 | XP_006253234.1 | 4 | 1020 | |||
XM_006253174.2 | XP_006253236.1 | 3 | 1056 | |||
XM_008771275.1 | XP_008769497.1 | 5 | 1055 | |||
XM_008771276.1 | XP_008769498.1 | 3 | 968 |
Gene Symbol: Zfp42l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237388.2 | XP_006237450.1 | 1 | 1456 |