Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106098.2 | NP_001099568.2 | 5 | 375 | ||
XM_006253493.2 | XP_006253555.1 | 5 | 1359 |