Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103690156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768432.1 | XP_008766654.1 | 1610 |
Gene Symbol: Galr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019172.4 | NP_062045.1 | 2 | 588 | ||
GenBank mRNA | AF008548.1 | - | 2 | 709 | ||
AF010318.1 | - | 2 | 588 | |||
U94322.1 | - | 2 | 1058 | |||
Y15248.1 | - | 2 | 568 |
Set Membership: |
G-protein coupled receptor |