Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alkbh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127450.1 | NP_001120922.1 | 5 | 346 | ||
GenBank mRNA | BC082048.1 | - | 5 | 346 | ||
BC098707.1 | - | 4 | 643 | |||
CB581428.1 | - | 5 | 352 |
Gene Symbol: LOC100912458 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228800.2 | XP_006228862.1 | 5 | 384 |