Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004222.1 | NP_001004222.1 | 4, 5 | 458 | ||
XM_006229933.2 | XP_006229995.1 | 4, 5 | 476 | |||
GenBank mRNA | BC079174.1 | - | 4, 5 | 458 | 7130075 |