Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024975.1 | NP_001020146.1 | 3 | 331 | ||
XM_008760402.1 | XP_008758624.1 | 5 | 512 | |||
GenBank mRNA | BC091345.1 | - | 3 | 331 | 7316315 | |
EF688599.1 | - | 3 | 331 |