Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199097.1 | NP_954528.1 | 2, 3 | 266 | ||
GenBank mRNA | AY346335.1 | - | 2, 3 | 266 | ||
BC092562.1 | - | 2, 3 | 255 | 7326245 |