Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd17a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006983.1 | NP_001006984.1 | 3 | 810 | ||
XM_008765090.1 | XP_008763312.1 | 3 | 3388 | |||
GenBank mRNA | BC083686.1 | - | 3 | 810 | 7189228 |