Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eme2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750789.3 | XP_003750837.1 | 3 | 499 | ||
XM_008767602.1 | XP_008765824.1 | 3 | 499 | |||
XM_008767603.1 | XP_008765825.1 | 3 | 502 | |||
XM_008767604.1 | XP_008765826.1 | 3 | 502 | |||
XM_008767605.1 | XP_008765827.1 | 3 | 465 | |||
GenBank mRNA | BC158841.1 | - | 3 | 445 |